The thickness is not determined for this product. In general, glass plates used for electrophoresis gels range from 0.125 to 0.1875 inches in thickness.
G0901
Glass plates for vertical standard gel
16.5 cm x 14.5 cm
About This Item
Recommended Products
Product Name
Glass plates for vertical standard gel, 16.5 cm wide, W × H 16.5 cm × 14.5 cm
W × H
16.5 cm × 14.5 cm
Packaging
Choose from one of the most recent versions:
Certificates of Analysis (COA)
-
How thick is the plate (in mm)?
1 answer-
Helpful?
-