Skip to Content
Merck
All Photos(1)

Key Documents

EP951033201

Eppendorf® Deepwell plates, DNA LoBind, 96 wells

white plate, conical bottom, colorless wells, capacity 1000 μL, pkg of 80 ea (10 bags × 8 plates)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
material:
clear wells
colorless wells
conical bottom
polypropylene
white plate
size:
96 well
sterility:
non-sterile
feature:
lid: no

material

clear wells
colorless wells
conical bottom
polypropylene
white plate

sterility

non-sterile

feature

lid: no

packaging

pkg of 80 ea (10 bags × 8 plates)

manufacturer/tradename

Eppendorf® 951033201

capacity

1000 μL

size

96 well

well volume

1000 μL

color

white border

suitability

suitable for PCR
suitable for molecular biology

Looking for similar products? Visit Product Comparison Guide

General description

Deepwell Plate 96/1000µL, DNA LoBind, wells clear, 1,000 µL, LoBind, PCR clean, white, 80 plates (10 bags × 8 plates)
  • Eppendorf LoBind material ensures optimized sample recovery for improved assay results
  • Free of surface coating (e.g. silicone) to minimize the risk of sample interference
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors
  • Available in tube, microplate, and deepwell plate formats for easy-up scaling
  • High-contrast Unique OptiTrack® matrix: up to 30 % faster sample identification and fewer pipetting errors
  • RecoverMax well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties
  • Raised well rims and a smooth surface ensure reliable sealing

Features and Benefits

  • Eppendorf LoBind material ensures optimized sample recovery for improved assay results
  • Free of surface coating (e.g. silicone) to minimize the risk of sample interference
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors
  • Available in tube, microplate, and deepwell plate formats for easy-up scaling
  • High-contrast Unique OptiTrack® matrix: up to 30 % faster sample identification and fewer pipetting errors
  • RecoverMax® well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties
  • Raised well rims and a smooth surface ensure reliable sealing

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
OptiTrack is a registered trademark of Eppendorf AG
RecoverMax is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nikos Tsolakos et al.
Vaccine, 28(18), 3211-3218 (2010-03-02)
In this study, we evaluated the effect of the growth medium on the composition and immunogenicity of meningococcal outer membrane vesicle (OMV) vaccines after cultivation of the Norwegian serogroup B 44/76 vaccine strain in either Frantz' or modified Catlin-6 media
E I Trilisky et al.
Journal of chromatography. A, 1142(1), 2-12 (2007-01-24)
Purified viruses are used in gene therapy and vaccine production. Ion-exchange chromatography (IEC) is the most common method for large-scale downstream purification of viruses and proteins. Published IEC protocols provide details for specific separations but not general methods for selecting
Sharon N Finger et al.
Nucleic acids research, 36(4), 1260-1272 (2008-01-05)
Telomerase is a ribonucleoprotein enzyme that maintains chromosome ends through de novo addition of telomeric DNA. The ability of telomerase to interact with its DNA substrate at sites outside its catalytic centre ('anchor sites') is important for its unique ability
M Yang et al.
International journal of pharmaceutics, 331(2), 176-181 (2006-11-28)
Salmon calcitonin (sCT) powders suitable for inhalation, containing chitosan and mannitol as absorption enhancer and protection agent, respectively, were prepared using a spray-drying process. The effect of chitosan on physicochemical stability of sCT in the dry powder was investigated by
Hartmut Stocker et al.
Antimicrobial agents and chemotherapy, 50(2), 667-673 (2006-01-27)
Therapeutic drug monitoring (TDM) is gaining importance for improving the success of antiretroviral treatment in human immunodeficiency virus-infected patients. However, enfuvirtide (ENF) concentrations are not regularly determined. The objective of this work was to study the pharmacokinetics (PK) of ENF

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service