Skip to Content
Merck
All Photos(1)

Key Documents

EP951020303

Eppendorf twin.tec® PCR Plates 96

semi skirted, size 250 μL, colorless, pkg of 25 ea

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NB.22
size:
250 μL
96 well
sterility:
non-sterile
feature:
skirt
semi skirted

sterility

non-sterile

feature

skirt
semi skirted

packaging

pkg of 25 ea

manufacturer/tradename

Eppendorf® 951020303

technique(s)

PCR: suitable

size

250 μL
96 well

color

colorless

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Choose Eppendorf twin.tec PCR plates to achieve reliable PCR results every time. twin.tec plates offer purity, stability, different colors and other simple, intelligent features that make your everyday work easier. The stable polycarbonate frame is well suited for automated and high-throughput processes, and the different colors allow you to quickly organize the individual steps of your workflow.

Application

Eppendorf twin.tec® PCR Plates 96 has been used in:
  • droplet digital PCR
  • DNA quantification and library preparation
  • single-cell sorting
PCR

Features and Benefits

  • The plate features a semi-skirted design, allowing space for labels and barcodes.
  • It is compatible with most thermal cyclers.
  • Has a maximum well volume of 250 µL (when used with cap strips).
  • The plate offers improved well-to-well tolerance.
  • Barcode functionality is available upon request, enabling convenient sample tracking and identification..
  • The plate is well-suited for quantitative real-time polymerase chain reaction (PCR)
  • The OptiTrack® matrix enhances sample identification speed and reduces pipetting errors.
  • Maximum recovery rates for improved assay results
  • Available with barcode (upon request)
  • Ideal for quantitative real-time PCR as well
  • Exceptionally solid and torque-resistant polycarbonate frame
  • OptiTrack® matrix for faster sample identification and fewer pipetting errors
  • Raised well rims for effective sealing, also reduces risk of cross-contamination
  • Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
OptiTrack is a registered trademark of Eppendorf AG
twin.tec is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Impact of BCG vaccination on the repertoire of human ?? T cell receptors.
Xia M, et al.
Frontiers in immunology, 14, 1100490-1100490 (2023)
Ali Naini et al.
Methods in cell biology, 155, 383-400 (2020-03-19)
The maternally inherited mitochondrial DNA (mtDNA) is a circular 16,569bp double stranded DNA that encodes 37 genes, 24 of which (2 rRNAs and 22 tRNAs) are necessary for transcription and translation of 13 polypeptides that are all subunits of respiratory
Noora Puhakka et al.
PloS one, 12(3), e0172521-e0172521 (2017-03-09)
Traumatic brain injury (TBI) can result in several dentate gyrus-regulated disabilities. Almost nothing is known about the chronic molecular changes after TBI, and their potential as treatment targets. We hypothesized that chronic transcriptional alterations after TBI are under microRNA (miRNA)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service