Recommended Products
manufacturer/tradename
Eppendorf® 6135010004
General description
Automatic print-out of results
Easy and direct connection via USB
Compact and light-weight (approx. 0.5 kg)
Easy paper operation
Max. printing speed: 90 mm/sec.
Number of dots/line: 832
Resolution (dots/mm): 8
Easy and direct connection via USB
Compact and light-weight (approx. 0.5 kg)
Easy paper operation
Max. printing speed: 90 mm/sec.
Number of dots/line: 832
Resolution (dots/mm): 8
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service