Key Documents
EP5391070026
Eppendorf(R) Plate Adapter HeatSealer S100
for 96 and 384 well plates
About This Item
UNSPSC Code:
41121815
NACRES:
NB.15
feature
for 96 and 384 well plates
manufacturer/tradename
Eppendorf® 5391070026 (Global)
Related Categories
Features and Benefits
Hermetic Sealing, of multiwell plates
Eliminates evaporation in PCR
Seals 96- and 384-well plates
Integrated thermostat prevents overheating
Improved mechanics for even easier Sealing,
PCR Plate Adapter included
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)