Key Documents
EP022638947
Adapter for Eppendorf® A-2-MTP Rotor
holds 1 x 96-well PCR plate, pack of 2 ea
About This Item
UNSPSC Code:
41103911
Pricing and availability is not currently available.
feature
holds 1 x 96-well PCR plate
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 022638947 (NA)
technique(s)
PCR: suitable
centrifugation: suitable
compatibility
for use with A-2-DWP-AT
for use with A-2-DWP
for use with A-4-62-MTP
for use with Rotors A-4-81 MTP/Flex
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)