Skip to Content
Merck
All Photos(1)

Documents

EP022491148

Eppendorf® epT.I.P.S. Biopur®,singles

volume 2-200 μL, L 53 mm, yellow

Synonym(s):

pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600

material

polypropylene
yellow

sterility

sterile

feature

DNase free
RNase free
endotoxin free

packaging

pack of 100 ea

manufacturer/tradename

Eppendorf® 022491148

L

53 mm

volume

2-200 μL

Looking for similar products? Visit Product Comparison Guide

General description

epT.I.P.S.® Singles, Biopur®, 2 – 200 µL, 53 mm, yellow, colorless tips, 100 tips, individually packed. Replaces order no. 0030010035.
  • Eppendorf Biopur pipette tip, individually wrapped
  • Ensured PCR clean, sterile, pyrogen-, ATP- and DNA-free
  • For use in pharmaceutical and food industry, molecular biology and cell technology
  • Batch number and expiration date on each blister package
  • Each production lot inspected continuously by an independent laboratory
  • Lot-specific certificates available at www.eppendorf.com/certificates

Features and Benefits

  • Eppendorf Biopur pipette tip, individually wrapped
  • Ensured PCR clean, sterile, pyrogen-, ATP- and DNA-free
  • For use in pharmaceutical and food industry, molecular biology and cell technology
  • Batch number and expiration date on each blister package
  • Each production lot inspected continuously by an independent laboratory
  • Lot-specific certificates available at www.eppendorf.com/certificates

Legal Information

Biopur is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service