Skip to Content
Merck
All Photos(1)

Key Documents

EHU100711

Sigma-Aldrich

MISSION® esiRNA

targeting human TGM2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AGAGCATGAACATGGGCAGTGACTTTGACGTCTTTGCCCACATCACCAACAACACCGCTGAGGAGTACGTCTGCCGCCTCCTGCTCTGTGCCCGCACCGTCAGCTACAATGGGATCTTGGGGCCCGAGTGTGGCACCAAGTACCTGCTCAACCTCAACCTGGAGCCTTTCTCTGAGAAGAGCGTTCCTCTTTGCATCCTCTATGAGAAATACCGTGACTGCCTTACGG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robin Delaine-Smith et al.
Cancers, 11(5) (2019-05-24)
Colorectal cancer is the third most common cancer worldwide, and the fourth leading cause of malignancy-related mortality. This highlights the need to understand the processes driving this disease in order to develop new treatments and improve patient outcomes. A potential
Yesim Bagatur et al.
Cell adhesion & migration, 12(2), 138-151 (2017-05-13)
Tissue transglutaminase (TG2) is the ubiquitously expressed member of transglutaminase family and shown to play a critical role in the development and progression of drug resistance malignancies. We have previously showed the association of TG2 upregulation with progression and metastasis
Sung-Yup Cho et al.
Life science alliance, 3(3) (2020-02-23)
Hypoxia selectively enhances mRNA translation despite suppressed mammalian target of rapamycin complex 1 activity, contributing to gene expression reprogramming that promotes metastasis and survival of cancer cells. Little is known about how this paradoxical control of translation occurs. Here, we
Emi Aonuma et al.
Biochemical and biophysical research communications, 542, 17-23 (2021-01-23)
Nickel, the most frequent contact allergy cause, is widely used for various metallic materials and medical devices. Autophagy is an intracellular protein degradation system and contributes to metal recycling. However, it is unclear the functions of nickel in autophagy. We
Deborah T Leicht et al.
Journal of thoracic oncology : official publication of the International Association for the Study of Lung Cancer, 9(6), 872-881 (2014-05-16)
Esophageal adenocarcinomas (EAC) are aggressive cancers that are increasing in incidence and associated with a poor prognosis. The identification of highly expressed genes in EAC relative to metaplastic Barrett's esophagus (BE) may provide new targets for novel early cancer detection

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service