Skip to Content
Merck
All Photos(1)

Key Documents

EHU078041

Sigma-Aldrich

MISSION® esiRNA

targeting human PMAIP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TTCTCAGGAGGTGCACGTTTCATCAATTTGAAGAAAGACTGCATTGTAATTGAGAGGAATGTGAAGGTGCATTCATGGGTGCCCTTGGAAACGGAAGATGGAATACATCAAAGTGAATTTCTGTTCAAGTTTTCCCAGATTATCATTCTTTGGGATGAGAGAACATTATAAAACCACTTTGTTTATTTTAAAGCAAGAATGGAAGACCCTTGAAAATAAAGAAGTAATTATTGACACATTTCTTTTTTACTTAGAGAATCGTTCTAGTGTTTTTGCCGAAGATTACCGCTGGCCTACTGTGAAGGGAGATGACCTGTGATTAGACTGGGCGGCTGGGGAGAAACAGTTCAGTGCATTGTTGTTGTTGCTGTTTTTGGTGTTTTGCTTTTCAGTGCCAACTCAGCAC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhenqian Wu et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 95, 1574-1579 (2017-09-28)
Colorectal cancer (CRC) cells undergo apoptosis in the presence of the small-molecule inhibitor ABT-263 by up-regulating antiapoptotic Bcl-2 family members. However, the resistance to ABT-263 gradually developed in most solid tumors due to its low affinity to Mcl-1. Here, we
Patricia Gomez-Bougie et al.
Cancer letters, 383(2), 204-211 (2016-10-25)
As myeloma cells actively produce and secrete immunoglobulins, they are prone to ER stress, which if unresolved leads to apoptosis. We found that myeloma cell death induced by the ER stressor Thapsigargin was highly variable, ranging from 2 to 89%.
Eugene Y Kim et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, fj201800425R-fj201800425R (2018-05-26)
Rheumatoid arthritis (RA) is characterized by hyperplastic pannus formation mediated by activated synovial fibroblasts (RASFs) that cause joint destruction. We have shown earlier that RASFs exhibit resistance to apoptosis, primarily as a result of enhanced expression of myeloid cell leukemia-1
Yohei Sugimoto et al.
Molecular cancer therapeutics, 19(10), 1992-2000 (2020-08-28)
Rhabdoid tumor is an aggressive, early childhood tumor. Biallelic inactivation of the SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily B member 1 (SMARCB1)/integrase interactor 1 (INI1) gene is the only common genetic feature in rhabdoid tumors. Loss of SMARCB1 function
Carolane Seiller et al.
Cell death & disease, 11(5), 316-316 (2020-05-07)
Multiple myeloma is a plasma cell malignancy that escapes from apoptosis by heterogeneously over-expressing anti-apoptotic BCL2 proteins. Myeloma cells with a t(11;14) translocation present a particular vulnerability to BCL2 inhibition while a majority of myeloma cells relies on MCL1 for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service