Skip to Content
Merck
All Photos(1)

Documents

EHU023131

Sigma-Aldrich

MISSION® esiRNA

targeting human LAMC2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TTCAGCTGTCCAGCTTGCTATAATCAAGTGAAGATTCAGATGGATCAGTTTATGCAGCAGCTTCAGAGAATGGAGGCCCTGATTTCAAAGGCTCAGGGTGGTGATGGAGTAGTACCTGATACAGAGCTGGAAGGCAGGATGCAGCAGGCTGAGCAGGCCCTTCAGGACATTCTGAGAGATGCCCAGATTTCAGAAGGTGCTAGCAGATCCCTTGGTCTCCAGTTGGCCAAGGTGAGGAGCCAAGAGAACAGCTACCAGAGCCGCCTGGATGACCTCAAGATGACTGTGGAAAGAGTTCGGGCTCTGGGAAGTCAGTACCAGAACCGAGTTCGGGATACTCACAGGCTCATCACTCAGATGCAGCTGAGCCTGGCAGAAAGTGAAGCTTCCTTGGGAAACACTAACATTCCTGCCTCAGACCACTACGTGGGGCCAAATGGCTTTAAAAGTCTGGCTCAGGAGGCCACAAGATTAGCAGAAAGCCACGTTGAGTCA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yao-Fei Pei et al.
The American journal of pathology, 189(8), 1637-1653 (2019-07-28)
Cholangiocarcinoma (CCA) is a malignant cancer that is associated with high mortality rates. The relationship between laminin γ 2 chain gene (LAMC2) and epidermal growth factor receptor (EGFR) has been previously documented in gastric cancer and oral squamous cell carcinoma.
Qiang-Hua Zhou et al.
Cancer management and research, 10, 2983-2995 (2018-09-15)
Molecular biomarkers, especially serologic factors, have been widely applied in cancer diagnosis and patient follow-up. However, there are few valuable prognostic factors in penile squamous cell carcinoma (PSCC). Here, the authors investigated whether laminin gamma 2 (LAMC2) expression, especially serum
D O Velez et al.
Nature communications, 8(1), 1651-1651 (2017-11-23)
The topographical organization of collagen within the tumor microenvironment has been implicated in modulating cancer cell migration and independently predicts progression to metastasis. Here, we show that collagen matrices with small pores and short fibers, but not Matrigel, trigger a
Manoj Garg et al.
Scientific reports, 7(1), 9749-9749 (2017-08-31)
Anaplastic thyroid carcinoma (ATC) is one of the most lethal malignancies having no effective treatment. Exportin-1 (XPO1) is the key mediator of nuclear export of many tumor suppressor proteins and is overexpressed in human cancers. In this study, we examined
Yan Liang et al.
Cell death and differentiation, 25(11), 1980-1995 (2018-03-08)
Esophageal squamous cell carcinoma (ESCC) is the main subtype of esophageal cancer. Long noncoding RNAs (lncRNAs) are thought to play a critical role in cancer development. Recently, lncRNA CASC9 was shown to be dysregulated in many cancer types, but the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service