CLS6795600D
Corning® hotplate with digital display
AC/DC input 120 V AC, US 3-pin plug, L × W 10 nL × 10 in.
Synonym(s):
stirrers
About This Item
Recommended Products
material
ceramic plate
packaging
case of 1
manufacturer/tradename
Corning 6795-600D
availability
not available in EU
parameter
5-550 °C temp. range
L × W
10 nL × 10 in.
Related Categories
General description
- Digital LED temperature display is adjustable in 5 °C increments and blinks until set temperature is reached
- Microprocessor maintains consistent and repeatable temperature settings.
- Bright LED HOT TOP icon lights up when top plate temperature is over 60 °C, even when heat control is turned off.
- Separate temperature sensor provides power cut off if unit overheats
- Extremely durable and heat resistant Pyroceram glass-ceramic top
- Small footprint and low profile
- Meets UL and cUL standards
- 2 year warranty
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)