Skip to Content
Merck
All Photos(2)

Key Documents

CLS352032

Corning® Falcon® Snap Caps

to hold, 12x75mm test tubes, sterile

Synonym(s):

Corning cap

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121700
NACRES:
NB.13
Pricing and availability is not currently available.

material

PE

sterility

sterile

feature

cap: no
to hold 12x75mm test tubes

manufacturer/tradename

Corning 352032

Application

Liquid Handling

Legal Information

Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service