CLS11750
Corning® closed system centrifuge tube
volume 500 mL, pre-assembled, sterile
Synonym(s):
500 mL centrifuge tubes, centrifuge tubes, polypropylene centrifuge tubes
About This Item
Recommended Products
material
cap (Plug Seal)
conical bottom tube
natural tube
polypropylene
sterility
sterile
feature
RCF 6,000 × g
packaging
pack of 2 ea
manufacturer/tradename
Corning 11750
parameter
6000 × g max. RCF
cap diam.
1.85 in.
tube O.D.
3.76 in.
volume
500 mL
color
clear
fitting
cap (Plug Seal)
General description
- Corning® closed system centrifuge tube is ideal for aseptic filling and transfer of samples to reduce the risk of contamination risk in liquid handling processes.
- The system is pre-assembled with the aseptic transfer cap to the centrifuge tube.
- The transfer caps have two ports: a venting port and a liquid transfer port. The venting port ends in a 0.2 μm filter, and the liquid transfer port, which is chemically resistant, heat sealable flexible tubing, ends in a male Luer.
- HDPE dip tubes facilitate easy aseptic transfer during your liquid handling processes.
- Tubes are sterile and nonpyrogenic.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service