Skip to Content
Merck
All Photos(1)

Key Documents

2H6-C2

95022203, Reactivity: Anti-Bovine Herpes Virus 1 (BHV-1)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Mouse Hybridoma

ImmunoDon

BALB/c mouse spleen

Myeloma

Sp2/0

Reactivity

Anti-Bovine Herpes Virus 1 (BHV-1)

technique(s)

cell culture | mammalian: suitable

isotype

IgG2b

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse Sp2/0 x Mouse BALB/c spleen

Cell Line Description

The antibody is directed against an immunodominant glycoprotein of bovine herpes 1 BHV-1 (infectious rhinotracheitis virus).

Immunogen

Bovine Herpes virus 1

Application

In vitro potency testing of BHV-1 vaccines.

Culture Medium

DMEM + 2mM glutamine + 2 5% Foetal Bovine Serum (FBS) + 5% Horse Serum (HS)

Subculture Routine

Maintain cultures between 100,000 - 1,000,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service