FHM
88102401, fish colon (posterior), Epithelial-like
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
product name
FHM, 88102401
biological source
fish colon (posterior)
growth mode
Adherent
karyotype
Modal no. 51
morphology
Epithelial-like
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Fish Fat Head minnow
Cell Line Description
Derived from tissue posterior to the anus of normal adult minnows. Reported to be capable of growth over a wide temperature range from 4°C-36°C with maximum growth at 34°C. Support growth of the virus of infectious pancreatic necrosis (IPN), cat fish tumour virus (CTV) and Granoff frog virus (FV1). ECHO-II will multiply in FHM but the cytopathic effect is not extensive.
Application
Virus studies: Inf. pacr. necrotic virus, cat fish tumour, and Granoff′s frog viruses
Culture Medium
EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) +10% Foetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:2 to 1:3; 34°C. Dislodge cells by agitation and scraping. Cells are reported to multiply over a wide range of temperatures 0°-36°C, with maximum growth at about 34°C. It is recommended that cultures are grown at 34
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service