This product is a cell line supplied by ECACC. The supplier notes that all cell lines are handled at containment level 2 in accordance with the Advisory Committee on Dangerous Pathogens (ACDP). This cell line was established in 1958, see J Exp Med 1958;108:945. For additional information please see the link below to review the supplier information:
https://www.culturecollections.org.uk/nop/product/cho
CHO Cell Line from Chinese hamster ovary
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 85050302, hamster ovary, Epithelial
Synonym(s):
CHO Cells, CHO-ori Cells, Chinese Hamster Ovary Cells, Chinese hamster ovary cell line
About This Item
Recommended Products
biological source
hamster ovary
packaging
tube of 5 μg 85050302-DNA-5UG
pkg of vial of cells 85050302-1VL
availability
not available in Japan
growth mode
Adherent
karyotype
Hypodiploid, modal no. 20
morphology
Epithelial
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Cell Line Description
Application
Culture Medium
Subculture Routine
Other Notes
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Eukaryotic cell lines aid in complex post-translational modifications and folding of recombinant proteins. Find suitable recombinant protein production cell lines.
-
Hi there, CHO cells can be used at BSL I lab, right? Best regards, Cristina
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service