Recommended Products
product name
CP5, 08052101
biological source
bovine connective tissue
growth mode
Adherent
karyotype
Not specified
morphology
Fibroblast
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Bos taurus (Holstein-Fresian) articular cartilage progenitor
Cell Line Description
A bovine (Bos taurus, Holstein-Fresian) articular cartilage progenitor cell line able to re-differentiate into articular chondrocytes after extensive population doublings (>30pd).
Culture Medium
DMEM:HAMS F12 (1:1) + 2mM Glutamine + 10% Foetal Bovine Serum (FBS) + 50ÂÂμg/ml ascorbate + 10mM HEPES
Subculture Routine
Split sub-confluent cultures (70-80%) 1:4 to 1:8 using 0.25% trypsin or trypsin/EDTA; 5% CO2;37ºC.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service