AFRC MAC 61.6
01010804, Reactivity: Anti-Abscisic acid, plant growth hormone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
biological source
Rat x Rat Hybridoma
ImmunoDon
F344 Rat spleen
Myeloma
YB2/3.0 Ag20
Reactivity
Anti-Abscisic acid, plant growth hormone
technique(s)
cell culture | mammalian: suitable
isotype
IgG2c
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Rat x Rat hybridoma
Cell Line Description
This cell line is the result of recloning of AFRC MAC 61. The antibody is directed against the plant growth hormone cis, trans-abscisic acid. The antibody production rate of this hybridoma has been tested at ECACC. It must be noted that this hybridoma produces less than 2μg/ml of unpurified antibody.
Immunogen
S-cis, trans-abscisic acid - 4(1)- p-amino-benzoyl-hydrazone-KLH conjugate
Culture Medium
Subculture Routine
Maintain cultures between 3-9 x 100,000 cell/ml; 5%CO2; 37°C. Require mouse macrophage feeder layers upon resuscitation
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service