Skip to Content
Merck
All Photos(1)

Key Documents

C9985

Sigma-Aldrich

Nalgene® Labtop Cooler

-20 °C labtop cooler, polycarbonate, blue, pack of 1

Synonym(s):

benchtop cooler, labtop cooler

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103025
NACRES:
NB.22
Pricing and availability is not currently available.

material

polycarbonate

packaging

pack of 1

manufacturer/tradename

Nunc 355501

L × W × H

226 mm (165 mm) × 133 mm

color

blue

Looking for similar products? Visit Product Comparison Guide

General description

Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using a benchtop cooler.

Features and Benefits

  • Holds 20 tubes, inserts are included to accommodate 0.2 to 0.5 mL microcentrifuge tubes.
  • Protects enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench (T<20 °C maintained up to 1 hour).
  • Maintains freezer temperature during the frost free cycle, power failure and accidental door opening.
  • Printed grid on one way keyed lid for proper orientation and sample identification.
  • Moulded of impact resistant polycarbonate; stackable, with locking handle.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service