C7855
Anti-Cdh1 antibody,Mouse monoclonal
clone DCS-266, purified from hybridoma cell culture
Synonym(s):
Anti-CDC20C, Anti-CDH1, Anti-FZR, Anti-FZR2, Anti-HCDH, Anti-HCDH1
About This Item
Recommended Products
biological source
mouse
conjugate
unconjugated
antibody form
purified from hybridoma cell culture
antibody product type
primary antibodies
clone
DCS-266, monoclonal
form
buffered aqueous solution
mol wt
antigen 50 kDa
species reactivity
human
concentration
~2 mg/mL
technique(s)
immunoprecipitation (IP): suitable
microarray: suitable
western blot: 2-4 μg/mL using a HeLa cell nuclear extract
isotype
IgG1
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... CDH1(999)
General description
Specificity
Immunogen
Application
Biochem/physiol Actions
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service