Skip to Content
Merck
All Photos(2)

Key Documents

C4528

Sigma-Aldrich

Chlorobimane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11ClN2O2
CAS Number:
Molecular Weight:
226.66
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

storage temp.

2-8°C

SMILES string

CC1=C(C)C(=O)N2N1C(CCl)=C(C)C2=O

InChI

1S/C10H11ClN2O2/c1-5-7(3)12-8(4-11)6(2)10(15)13(12)9(5)14/h4H2,1-3H3

InChI key

SUIPVTCEECPFIB-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

María Cristina Estañ et al.
PloS one, 9(12), e115250-e115250 (2014-12-17)
Fatty acid synthesis and oxidation are frequently exacerbated in leukemia cells, and may therefore represent a target for therapeutic intervention. In this work we analyzed the apoptotic and chemo-sensitizing action of the fatty acid oxidation inhibitor etomoxir in human acute
Emma Dreischmeier et al.
Analytical biochemistry, 616, 114100-114100 (2021-01-09)
PrC-210 is a direct-acting ROS-scavenger. It's active when administered orally, IV, or topically; it has none of the nausea/emesis nor hypotension side effects that have precluded human amifostine use. PrC-210 confers 100% survival to mice and rats that received an
Pandian Nagakannan et al.
Biochimica et biophysica acta. Molecular cell research, 1868(3), 118928-118928 (2020-12-20)
Ferroptosis is a necrotic form of cell death caused by inactivation of the glutathione system and uncontrolled iron-mediated lipid peroxidation. Increasing evidence implicates ferroptosis in a wide range of diseases from neurotrauma to cancer, highlighting the importance of identifying an
Seoungwoo Shin et al.
International journal of molecular sciences, 15(10), 18919-18940 (2014-10-22)
The phenolic compound phloretin is a prominent member of the chemical class of dihydrochalcones. Phloretin is specifically found in apple and apple juice and known for its biological properties. We were particularly interested in its potential dermo-cosmetic applications. However, practical
Elsa Gouix et al.
Pharmacological research, 87, 8-17 (2014-06-15)
Understanding the role of astrocytes in stroke is assuming increasing prominence, not only as an important component on its own within the neurovascular unit, but also because astrocytes can influence neuronal outcome. Ischemia may induce astrogliosis and other phenotypic changes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service