Skip to Content
Merck
All Photos(1)

Key Documents

C3236

Sigma-Aldrich

Nunc® CryoTubes® storage cane

storage cane for cryotubes, aluminum, 50/cs

Synonym(s):

Cyrotube storage device, cryostorage cane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
Pricing and availability is not currently available.

material

aluminum

packaging

case of 50
pack of 50

manufacturer/tradename

Nunc 378441

General description

Standard Nunc® aluminim cryo storage cane, 11.31 in (29 cm) long, pack of 50.

Legal Information

CryoTube is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service