Yes, the final concentration of BPE is 0.004 mg/mL.
C-20011
Keratinocyte Growth Medium 2
Ready-to-use kit including Basal Medium and SupplementMix, 500 ml
Synonym(s):
Primary Cell Culture Media
About This Item
Recommended Products
form
liquid
packaging
pkg of 500 ml
technique(s)
cell culture | mammalian: suitable
shipped in
wet ice
General description
Application
Components
Quality
Preparation Note
Storage and Stability
Subculture Routine
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Primary human keratinocytes and serum-free keratinocyte growth media for the feeder cell-free in vitro cultivation of epidermal keratinocytes. Protocols for handling, cell passaging, media, and product use.
Related Content
Explore expert protocols and culture media for human primary cell lines to replicate biological systems accurately.
Explore expert protocols and culture media for human primary cell lines to replicate biological systems accurately.
Explore expert protocols and culture media for human primary cell lines to replicate biological systems accurately.
Explore expert protocols and culture media for human primary cell lines to replicate biological systems accurately.
-
what is the concentration of BPE? Is it 0.004 mg/ml ?.
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service