Skip to Content
Merck
All Photos(1)

Key Documents

B5063

Sigma-Aldrich

BMS-193885

≥98% (HPLC)

Synonym(s):

1,4-dihydro-4-[3-[[[[3-[4-(3-methoxyphenyl)-1-piperidinyl]propyl]amino]carbonyl]amino]phenyl]-2,6-dimethyl-3,5-dimethyl ester-3,5-pyridinedicarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H42N4O6
CAS Number:
Molecular Weight:
590.71
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

light yellow to yellow

solubility

DMSO: >10 mg/mL

originator

Bristol-Myers Squibb

storage temp.

−20°C

SMILES string

COC(=O)C1=C(C)NC(C)=C(C1c2cccc(NC(=O)NCCCN3CCC(CC3)c4cccc(OC)c4)c2)C(=O)OC

InChI

1S/C33H42N4O6/c1-21-28(31(38)42-4)30(29(22(2)35-21)32(39)43-5)25-10-6-11-26(19-25)36-33(40)34-15-8-16-37-17-13-23(14-18-37)24-9-7-12-27(20-24)41-3/h6-7,9-12,19-20,23,30,35H,8,13-18H2,1-5H3,(H2,34,36,40)

InChI key

WMYSXJSJXZFODY-UHFFFAOYSA-N

Biochem/physiol Actions

BMS-193885 is a potent, selective Y1 antagonist that is active in both acute and chronic animal models of food intake.
BMS-193885 is a potent, selective Y1 antagonist that is active in both acute and chronic animal models of food intake. Although it is active in vivo, it is not orally bioavailable due to poor intestinal absorption, so it is not being pursued for pharmaceutical development. BMS-193885 has been used as a pre-clinical proof of concept tool for showing efficacy of Y1 antagonism in treating obesity.

Features and Benefits

This compound is featured on the Neuropeptide Y Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Bristol-Myers Squibb. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shiba Yousefvand et al.
Canadian journal of physiology and pharmacology, 96(12), 1301-1307 (2018-10-17)
Neuropeptide Y (NPY) plays a mediatory role in cerebral insulin function by maintaining energy balance. The current study was designed to determine the role of insulin in food intake and its interaction with NPY receptors in 8 experiments using broiler
David Acton et al.
Cell reports, 28(3), 625-639 (2019-07-18)
Acute itch can be generated by either chemical or mechanical stimuli, which activate separate pathways in the periphery and spinal cord. While substantial progress has been made in mapping the transmission pathway for chemical itch, the central pathway for mechanical

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service