Skip to Content
Merck
All Photos(1)

Key Documents

AXYSCT50ML500

Centrifuge tube with screw cap, conical bottom, co-polymer

capacity 50 mL, non-sterile

Synonym(s):

15 ml centrifuge tubes, polypropylene centrifuge tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121703
NACRES:
NB.11

material

conical bottom tube
polypropylene
screw top cap

Agency

suitable for EPA 200.7
suitable for EPA 200.8

sterility

non-sterile

packaging

pack of 50 ea
case of 500 ea

manufacturer/tradename

Axygen Inc SCT-50ML-500
Corning

parameter

12000 × g max. RCF

capacity

50 mL

color

clear

fitting

cap (screw cap)

suitability

suitable for (centrifugation)

Looking for similar products? Visit Product Comparison Guide

General description

  • Axygen® centrifuge tubes are made of chemically resistant polypropylene.
  • Features black printed graduations for precise volume measurement
  • Large white marking spot for sample identification.

Legal Information

Axygen is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service