Skip to Content
Merck
All Photos(1)

Key Documents

AXYP2MLSQC

Deep well plate

96 square wells, working volume 2.0 mL, 25 ea of

Synonym(s):

Corning cell culture plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
material:
square bottom clear polypropylene wells
maximum volume:
2.2 mL
size:
96 wells
working volume:
2.0 mL
Pricing and availability is not currently available.

material

square bottom clear polypropylene wells

sterility

non-sterile

feature

lid: no
skirt

packaging

case of 25 ea
pack of 5 ea
25 ea of

manufacturer/tradename

Axygen Inc P-2ML-SQ-C
Corning

maximum volume

2.2 mL

size

96 wells

working volume

2.0 mL

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Axygen® deep well and assay microplates are made of chemically resistant and temperature resistant high-quality polypropylene.
  • Ideal for sample collection, invitro growth chamber, and long-term storage.
  • The square-well design of microplate ensures complete sample recovery.
  • The plates can be sealed using a heat-sealing film for sample protection.

Legal Information

Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service