Skip to Content
Merck
All Photos(1)

Key Documents

AV46950

Sigma-Aldrich

Anti-ST6GALNAC6 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-RP11-203J24.3, Anti-SIAT7F, Anti-ST6 -N-acetylgalactosaminide α-2,6-sialyltransferase 6, Anti-ST6GALNACVI

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
WB
clone:
polyclonal
species reactivity:
rabbit, dog, guinea pig, horse, human, rat, mouse
citations:
2
technique(s):
western blot: suitable
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

38 kDa

species reactivity

rabbit, dog, guinea pig, horse, human, rat, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

Synthetic peptide directed towards the C terminal region of human ST6GALNAC6

Application

Anti-ST6GALNAC6 antibody produced in rabbit is suitable for western blotting at a concentration of 0.5μg/mL.

Biochem/physiol Actions

ST6GALNAC6 [ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 6] gene encodes a single-pass type II membrane protein that belongs to glycosyltransferase 29 family and is primarily expressed in kidney, in proximal tubule epithelial cells and colon cell lines. It plays a crucial role in biosynthesis of DSGG (disialylgalactosylgloboside) from MSGG (monosialylgalactosylgloboside) in normal and malignant kidney. It is also involved in the synthesis of disialyl Lewis a (Lea).

Sequence

Synthetic peptide located within the following region: YHYYEPKGPDECVTYIQNEHSRKGNHHRFITEKRVFSSWAQLYGITFSHP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akiko Tsuchida et al.
The Journal of biological chemistry, 278(25), 22787-22794 (2003-04-02)
Biosynthesis of disialyl Lewis a (Lea) was analyzed using previously cloned ST6GalNAc V and ST6GalNAc VI, which were responsible for the synthesis of alpha-series gangliosides. Among lactotetraosylceramide (Lc4), neolactotetraosylceramide, and their sialyl forms, only sialyl Lc4 was sialylated with ST6GalNAc
Motohiro Senda et al.
The Biochemical journal, 402(3), 459-470 (2006-11-25)
Although disialyl glycosphingolipids such as GD3 and GD2 have been considered to be associated with malignant tumours, whether branched-type disialyl glycosphingolipids show such an association is not well understood. We investigated the sialyltransferases responsible for the biosynthesis of DSGG (disialylgalactosylgloboside)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service