Skip to Content
Merck
All Photos(3)

Key Documents

AV32519

Sigma-Aldrich

Anti-FOSB antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-FBJ murine osteosarcoma viral oncogene homolog B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

36 kDa

species reactivity

human, horse, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FOSB(2354)

General description

FOSB is a leucine zipper protein that functions as a transcriptional regulator. Studies in mice have reported that FOSB is involved in cocaine-induced behavioral responses.
Rabbit Anti-FOSB antibody recognizes human, mouse, rat, bovine, and canine FOSB.

Immunogen

Synthetic peptide directed towards the C terminal region of human FOSB

Application

Rabbit Anti-FOSB antibody can be used for IHC (4-8μg/ml) and western blot (1.25μg/ml) applications.

Biochem/physiol Actions

The Fos gene family consists of 4 members: FOS, FOSB, FOSL1, and FOSL2. They are leucine zipper proteins that can dimerize with proteins of the JUN family, thereby forming the transcription factor complex AP-1. As such, the FOS proteins have been implicated as regulators of cell proliferation, differentiation, and transformation.

Sequence

Synthetic peptide located within the following region: DLPGSAPAKEDGFSWLLPPPPPPPLPFQTSQDAPPNLTASLFTHSEVQVL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R F De Pauli et al.
Pharmacology, biochemistry, and behavior, 117, 70-78 (2013-12-21)
Chronic drug exposure and drug withdrawal induce expressive neuronal plasticity which could be considered as both functional and pathological responses. It is well established that neuronal plasticity in the limbic system plays a pivotal role in relapse as well as
Josy Carolina C Pontes et al.
Epilepsy research, 126, 16-25 (2016-07-16)
The efficiency of most of the new antiepileptic drugs (AEDs) on clinical trials still falls short the success reported in pre-clinical studies, possibly because the validity of the animal models is insufficient to fully represent the human pathology. To improve
M C Gruda et al.
Oncogene, 12(10), 2177-2185 (1996-05-16)
FosB, one of the members of the Fos family, is rapidly induced in many cell types upon stimulation and has a stimulatory effect on the proliferation of cultured cells. To understand the tissue distribution of FosB, we have studied its
N Hiroi et al.
Proceedings of the National Academy of Sciences of the United States of America, 94(19), 10397-10402 (1997-09-18)
Chronic exposure to cocaine leads to prominent, long-lasting changes in behavior that characterize a state of addiction. The striatum, including the nucleus accumbens and caudoputamen, is an important substrate for these actions. We previously have shown that long-lasting Fos-related proteins
Izabelle Dias Benfato et al.
Behavioural brain research, 417, 113630-113630 (2021-10-18)
Social isolation gained discussion momentum due to the COVID-19 pandemic. Whereas many studies address the effects of long-term social isolation in post-weaning and adolescence and for periods ranging from 4 to 12 weeks, little is known about the repercussions of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service