A211150
DMT-2′O-Methyl-rA(bz) Phosphoramidite
configured for (ÄKTA® and OligoPilot®), configured for ABI
Synonym(s):
DMT-2′-O-Me-rA(bz) amidite, N-benzoyl-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-O-methyl-adenosine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite]
About This Item
Recommended Products
biological source
non-animal source (no BSE/TSE risk)
Quality Level
product line
Proligo Reagents
Assay
≥99% (31P-NMR)
≥99.0% (reversed phase HPLC)
form
powder
technique(s)
oligo synthesis: suitable
impurities
≤0.1% single unspecified Impurity (reversed phase HPLC)
≤0.3% mA2 (reversed phase HPLC, Hydrolysate)
≤0.3% mA3 (reversed phase HPLC, DMT-rA(bz)me)
≤0.3% water content (Karl Fischer)
≤0.5% P(III) Impurities 100-169ppm (31P-NMR)
≤1.0% mA1 (reversed phase HPLC, DMT-rA(bz)me-DMT)
≤3% residual Solvent content
color
white to off-white
λ
conforms (UV/VIS Identity)
suitability
conforms to structure for H-NMR
conforms to structure for LC-MS
compatibility
configured for (ÄKTA® and OligoPilot®)
configured for ABI
nucleoside profile
base: adenosine
base protecting group: benzoyl
2' protecting group: methyl
5' protecting group: DMT
deprotection: standard
storage temp.
2-8°C
SMILES string
CO[C@@H]1[C@H](OP(OCCC#N)N(C(C)C)C(C)C)[C@@H](COC(c2ccccc2)(c3ccc(OC)cc3)c4ccc(OC)cc4)O[C@H]1n5cnc6c(NC(=O)c7ccccc7)ncnc56
InChI
1S/C48H54N7O8P/c1-32(2)55(33(3)4)64(61-28-14-27-49)63-42-40(62-47(43(42)59-7)54-31-52-41-44(50-30-51-45(41)54)53-46(56)34-15-10-8-11-16-34)29-60-48(35-17-12-9-13-18-35,36-19-23-38(57-5)24-20-36)37-21-25-39(58-6)26-22-37/h8-13,15-26,30-33,40,42-43,47H,14,28-29H2,1-7H3,(H,50,51,53,56)/t40-,42-,43-,47-,64?/m1/s1
InChI key
AZCGOTUYEPXHMJ-PSVHYZMASA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
- Diagnostic probes
- Aptamer and ribozyme development
- Mixed 2′O-Methyl-RNA/DNA antisense molecules (a)
Legal Information
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service