Skip to Content
Merck
All Photos(1)

Key Documents

A1104

Sigma-Aldrich

Anti-Angiopoietin-1 antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-ANG-1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

goat

Quality Level

recombinant

expressed in goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

immunohistochemistry: 5-15 μg/mL using cells and tissues
indirect ELISA: 0.5-1.0 μg/mL
western blot: 0.1-0.2 μg/mL

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ANGPT1(284)

General description

Angiopoietin 1 is an endothelial cell-specific protein that is a ligand for the TIE2 tyrosine receptor. Angiopoietin 1 is critical for mature blood vessel formation.
Angiopoietin-1 (ANG-1) activates Tie-2 signaling in endothelial cells to promote chemotaxis, cell survival, cell sprouting, vessel growth, and stabilization,. ANG-1 is closely related to ANG-2. Both ANG-1 and ANG-2, with their receptor Tie-2, regulate embryonic vasculogenesis and angiogenisis, adult angiogenic sprouting, as well as endothelial cell proliferation.

Immunogen

purified, NSO-derived, recombinant human ANG-1.

Application

Goat anti-angiopoietin-1 can be used for immunohistochemistry (5-15 μg/mL), indirect ELISA (0.5-1.0 μg/mL) and western blot (0.1-0.2 μg/mL).

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Jones et al.
Nature reviews. Molecular cell biology, 2(4), 257-267 (2001-04-03)
Angiogenesis is required for normal embryonic vascular development and aberrant angiogenesis contributes to several diseases, including cancer, diabetes and tissue ischaemia. What are the molecular mechanisms that regulate this important process? The Tie family of receptors and their ligands, the
L M Orre et al.
Oncogene, 32(49), 5531-5540 (2013-06-12)
S100A4 is a small calcium-binding protein that is commonly overexpressed in a range of different tumor types, and it is widely accepted that S100A4 has an important role in the process of cancer metastasis. In vitro binding assays has shown
Shigetomo Fukuhara et al.
Histology and histopathology, 25(3), 387-396 (2010-01-08)
Angiopoietin (Ang) 1 is a ligand for endothelium-specific receptor tyrosine kinase Tie-2. In adult vasculature, Ang1/Tie2 signaling is thought to regulate both maintenance of vascular quiescence and promotion of angiogenesis. However, it has been unknown how Tie2 signal regulates these
B Witzenbichler et al.
The Journal of biological chemistry, 273(29), 18514-18521 (1998-07-11)
Angiopoietin-1 and its putative natural antagonist, angiopoietin-2, were recently isolated, and the critical role of angiopoietin-1 in embryogenic angiogenesis was demonstrated by targeted gene disruption. Specific biological effects of angiopoietin-1, however, have yet to be defined. In this study we
P C Maisonpierre et al.
Science (New York, N.Y.), 277(5322), 55-60 (1997-07-04)
Angiogenesis is thought to depend on a precise balance of positive and negative regulation. Angiopoietin-1 (Ang1) is an angiogenic factor that signals through the endothelial cell-specific Tie2 receptor tyrosine kinase. Like vascular endothelial growth factor, Ang1 is essential for normal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service