Skip to Content
Merck
All Photos(1)

Key Documents

95843

Sigma-Aldrich

Potassium acetate solution

BioUltra, for molecular biology, 5 M in H2O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOK
CAS Number:
Molecular Weight:
98.14
Beilstein:
3595449
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

grade

for molecular biology

Quality Level

product line

BioUltra

form

liquid

concentration

5 M in H2O

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected

pH

7.0-9.0 (25 °C)

density

1.215 g/mL at 20 °C

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤5 mg/kg

cation traces

Al: ≤1 mg/kg
As: ≤0.1 mg/kg
Ba: ≤1 mg/kg
Bi: ≤1 mg/kg
Ca: ≤20 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
Li: ≤1 mg/kg
Mg: ≤5 mg/kg
Mn: ≤1 mg/kg
Mo: ≤1 mg/kg
Na: ≤1000 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Sr: ≤1 mg/kg
Zn: ≤1 mg/kg

UV absorption

λ: 260 nm Amax: 0.04
λ: 280 nm Amax: 0.02

SMILES string

[K+].CC([O-])=O

InChI

1S/C2H4O2.K/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

SCVFZCLFOSHCOH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Potassium acetate 5M solution may be used as a stock to formulate precipitant solutions used in procedures such as the hanging-drop vapor diffusion crystallization protocol. Potassium acetate 5M solution may be used to prepare hypoosmotic media and in various DNA extraction protocols.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trevor R Sorrells et al.
eLife, 7 (2018-09-11)
Convergent evolutionary events in independent lineages provide an opportunity to understand why evolution favors certain outcomes over others. We studied such a case where a large set of genes-those coding for the ribosomal proteins-gained cis-regulatory sequences for a particular transcription
Michal Lubas et al.
EMBO reports, 19(6) (2018-05-02)
Autophagy is an essential catabolic process responsible for recycling of intracellular material and preserving cellular fidelity. Key to the autophagy pathway is the ubiquitin-like conjugation system mediating lipidation of Atg8 proteins and their anchoring to autophagosomal membranes. While regulation of
Hsiang-Tsui Wang et al.
Oncotarget, 7(49), 80450-80464 (2016-10-16)
Acrolein (Acr) is a potent cytotoxic and DNA damaging agent which is ubiquitous in the environment and abundant in tobacco smoke. Acr is also an active cytotoxic metabolite of the anti-cancer drugs cyclophosphamide and ifosfamide. The mechanisms via which Acr
Manuela Barilari et al.
The EMBO journal, 36(6), 736-750 (2017-03-01)
The inactivation of S6 kinases mimics several aspects of caloric restriction, including small body size, increased insulin sensitivity and longevity. However, the impact of S6 kinase activity on cellular senescence remains to be established. Here, we show that the constitutive
Eric J Wolf et al.
Cell reports, 31(8), 107693-107693 (2020-05-28)
The mammalian mRNA nuclear export process is thought to terminate at the cytoplasmic face of the nuclear pore complex through ribonucleoprotein remodeling. We conduct a stringent affinity-purification mass-spectrometry-based screen of the physical interactions of human RNA-binding E3 ubiquitin ligases. The

Questions

  1. What is the pH of this solution 95843 Potassium Acetate 5M

    1 answer
    1. The pH of this product may range from 7.0-9.0 at 25 °C. This information is lot specific and reported in the product Certificate of Analysis. Please see the link below to review a sample or lot specific Certificate:
      https://www.sigmaaldrich.com/product/sigma/95843#product-documentation

      Please see the link below to review the product datasheet:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/776/995/95843dat.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service