Skip to Content
Merck
All Photos(2)

Documents

62070

Sigma-Aldrich

Leupeptin

>4000 U/mg

Synonym(s):

Leupeptin hemisulfate salt, Acetyl-Leu-Leu-Arg-al, N-Acetyl-L-leucyl-L-leucyl-L-argininal hemisulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C20H38N6O4 · 1/2H2SO4
CAS Number:
Molecular Weight:
475.59
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic

form

powder

specific activity

>4000 U/mg

solubility

H2O: 50 mg/mL

storage temp.

−20°C

SMILES string

OS(O)(=O)=O.CC(C)C[C@H](NC(C)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCNC(N)=N)C=O.CC(C)C[C@H](NC(C)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCNC(N)=N)C=O

InChI

1S/2C20H38N6O4.H2O4S/c2*1-12(2)9-16(24-14(5)28)19(30)26-17(10-13(3)4)18(29)25-15(11-27)7-6-8-23-20(21)22;1-5(2,3)4/h2*11-13,15-17H,6-10H2,1-5H3,(H,24,28)(H,25,29)(H,26,30)(H4,21,22,23);(H2,1,2,3,4)/t2*15-,16-,17-;/m00./s1

InChI key

CIPMKIHUGVGQTG-VFFZMTJFSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Inhibitor of serine and cysteine proteases. Inhibits plasmin, trypsin, papain, calpain, and cathepsin B. Does not inhibit pepsin, cathepsins A and D, thrombin, or α-chymotrypsin. Effective concentration 10-100 μM. There have been numerous studies using leupeptin to protect against hearing loss caused by acoustic overstimulation or the ototoxic antibiotic gentamicin. (Loss of cochlear hair cells is believed to be mediated by calpain.)

Unit Definition

1 U corresponds to the amount of inhibitor which reduces the trypsin activity by 1 BAEE-U. (1 BAEE-U is the amount of enzyme which increases the absorbance at 253 nm by 0.001 per minute at pH 7.6 and 25 °C; BAEE, Cat. No. 12880, as substrate)

Analysis Note

Leupeptin gives multiple peaks on HPLC due to equilibria among three forms in solution. Purity determined using three main peaks. Majority of contaminating peptide is racemized leupeptin.

Other Notes

Inhibitor of trypsin, plasmin, papain and cathepsin B

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leupeptins, new protease inhibitors from Actinomycetes.
T Aoyagi et al.
The Journal of antibiotics, 22(6), 283-286 (1969-06-01)
Biological activities of leupeptins.
T Aoyagi et al.
The Journal of antibiotics, 22(11), 558-568 (1969-11-01)
A Kamal et al.
The Journal of cell biology, 142(4), 937-947 (1998-08-29)
Previously we reported that annexin VI is required for the budding of clathrin-coated pits from human fibroblast plasma membranes in vitro. Here we show that annexin VI bound to the NH2-terminal 28-kD portion of membrane spectrin is as effective as
Structures and activities of protease inhibitors of microbial origin.
H Umezawa
Methods in enzymology, 45, 678-695 (1976-01-01)
Yunzhong Zhang et al.
Journal of cellular physiology, 236(2), 1345-1361 (2020-07-14)
Fibroblast growth factor 21 (FGF21) as a metabolic stress hormone, is mainly secreted by the liver. In addition to its well-defined roles in energy homeostasis, FGF21 has been shown to promote remyelination after injury in the central nervous system. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service