Skip to Content
Merck
All Photos(1)

Key Documents

262A-1

Sigma-Aldrich

Granzyme B Rabbit Polyclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

Ig fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (262A-14)
vial of 0.5 mL concentrate (262A-15)
bottle of 1.0 mL predilute (262A-17)
vial of 1.0 mL concentrate (262A-16)
bottle of 7.0 mL predilute (262A-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:10-1:50

control

ALCL, spleen

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... GZMB(3002)

General description

Granzymes are serine proteases which are stored in specialized lytic granules of cytotoxic T-lymphocytes and in natural killer cells. Anti-granzyme B has been useful in diagnosing natural killer/T-cell lymphoma, as well as anaplastic large cell lymphoma. High percentages of cytotoxic T-cells have been shown to be an unfavorable indicator in Hodgkin disease.

Quality


IVD

IVD

IVD

RUO

Linkage

Granzyme B Positive Control Slides, Product No. 262S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J J Oudejans et al.
Blood, 89(4), 1376-1382 (1997-02-15)
Although the results of treatment of Hodgkin's disease (HD) have improved considerably in the last decades, the disease remains fatal in a minority of patients. We have recently shown that numbers of activated cytotoxic T cells (CTLs), present in tumor
J A Kummer et al.
Clinical and experimental immunology, 100(1), 164-172 (1995-04-01)
Cytoplasmic granules from activated natural killer (NK) and cytotoxic T lymphocytes (CTL) contain a pore-forming protein, perforin, and several homologous serine proteinases called granzymes. Expression of these proteins correlates with the cytolytic potential of cytotoxic lymphocytes. Using a panel of
Naoko Kato et al.
The American Journal of dermatopathology, 25(2), 142-147 (2003-03-26)
A neutrophil-rich anaplastic large cell lymphoma (ALCL) presented in the skin of a 47-year-old Japanese woman. The solitary cutaneous lesion was an eroded, 10-mm, dome-shaped nodule involving the skin of her left upper arm. Histologically, it showed a proliferation of
J J Oudejans et al.
The American journal of pathology, 148(1), 233-240 (1996-01-01)
Reed-Sternberg (RS) and Hodgkin's (H) cells are considered to be the neoplastic cells in Hodgkin's disease. Although most data suggest a lymphoid origin, the nature of these cells still remains the subject of considerable controversy. Recently, monoclonal antibodies became available
Jie Liu et al.
The Journal of dermatology, 30(10), 735-741 (2003-12-20)
Nasal natural killer (NK)/T cell lymphoma is an Epstein-Barr virus (EBV) associated lymphoma that arises in the nasal area and aggressively invades surrounding tissues. Our patient was a 48-year-old male who had had nasal obstruction and nasal discharge for 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service