Skip to Content
Merck
All Photos(1)

Key Documents

241R-4

Sigma-Aldrich

Cyclin D1 (EP12) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

EP12, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (241R-44)
vial of 0.5 mL concentrate (241R-45)
bottle of 1.0 mL predilute (241R-47)
vial of 1 mL concentrate (241R-46)
bottle of 7.0 mL predilute (241R-48)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

dilution

1:100-1:500

isotype

IgG

control

mantle cell lymphoma

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

Gene Information

human ... CCND1(595)

General description

Cyclin D1, one of the key cell cycle regulators, is a putative proto-oncogene overexpressed in a wide variety of human neoplasms. Cyclins are proteins that govern transitions through distinct phases of the cell cycle by regulating the activity of the cyclin-dependent kinases. In mid-to-late G1 phase of the cell cycle, cyclin D1 shows a maximum expression following growth factor stimulation. Anti-cyclin D1 has been successfully employed and is a promising tool for further studies in both cell cycle biology and cancer associated abnormalities. This antibody is useful for separating mantle cell lymphomas (cyclin D1 positive) from chronic lymphocytic leukemia/small lymphocytic lymphoma and follicular lymphomas (cyclin D1 negative). Hairy cell leukemia and plasma cell myeloma can weakly express Cyclin D1.

Quality


IVD

IVD

IVD

RUO

Linkage

Cyclin D1 Positive Control Slides, Product No. 241S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

WHO Classification of Tumours of Haematopoietic and Lymphoid Tissue, 189-189 (2008)
J Bartkova et al.
Cancer research, 55(4), 949-956 (1995-02-15)
D-type cyclins are proto-oncogenic cell cycle regulators implicated in the pathogenesis of several types of cancer. Amplification of the cyclin D1 gene has been described in 30-50% of human head and neck squamous cell carcinoma (HNSCC). Using immunohistochemistry on archival
R C Hankin et al.
Archives of pathology & laboratory medicine, 123(12), 1182-1188 (1999-12-03)
This article summarizes the most useful ancillary immunohistochemical and molecular assays for use in the diagnosis of mantle cell lymphoma. The English language literature was surveyed, with an emphasis on recent publications, for articles presenting key advances in the molecular
Roman Kodet et al.
Virchows Archiv : an international journal of pathology, 442(6), 538-547 (2003-05-03)
Mantle cell lymphoma (MCL) is a clinicopathological entity characterized by an aggressive clinical course, morphological features, and overexpression of cyclin D1 due to juxtaposition of the bcl-1 locus (and CCND1 gene coding for the cyclin D1) to the IgH gene.
J Bartkova et al.
Oncogene, 10(4), 775-778 (1995-02-16)
Cyclin D1 is a cell cycle regulator essential for G1 phase progression and a candidate proto-oncogene whose deregulated expression has been implicated in pathogenesis of several types of cancer. We have examined expression of cyclin D1 in 212 primary tumours

Related Content

Diagnostic immunohistochemistry overview highlights its importance in cancer diagnosis and detecting infectious diseases in modern clinical pathology.

Diagnostic immunohistochemistry overview highlights its importance in cancer diagnosis and detecting infectious diseases in modern clinical pathology.

Diagnostic immunohistochemistry overview highlights its importance in cancer diagnosis and detecting infectious diseases in modern clinical pathology.

Diagnostic immunohistochemistry overview highlights its importance in cancer diagnosis and detecting infectious diseases in modern clinical pathology.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service