Skip to Content
Merck
All Photos(1)

Key Documents

480967

Sigma-Aldrich

Sodium molybdate dihydrate

99.99% trace metals basis

Synonym(s):

Molybdic acid sodium salt dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2MoO4 · 2H2O
CAS Number:
Molecular Weight:
241.95
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
Assay:
99.99% trace metals basis
form:
powder or crystals
Pricing and availability is not currently available.

Quality Level

Assay

99.99% trace metals basis

form

powder or crystals

reaction suitability

reagent type: catalyst
core: molybdenum

pH

7-10.5 (25 °C, 5% in water)

mp

100 °C (dec.) (lit.)

density

2.37 g/mL at 25 °C

anion traces

chloride (Cl-): ≤0.005%
phosphate (PO43-): ≤5 ppm
sulfate (SO42-): ≤0.015%

cation traces

Fe: ≤0.001%
NH4+: ≤0.001%
heavy metals (as Pb): ≤5 ppm

SMILES string

O.O.[Na+].[Na+].[O-][Mo]([O-])(=O)=O

InChI

1S/Mo.2Na.2H2O.4O/h;;;2*1H2;;;;/q;2*+1;;;;;2*-1

InChI key

FDEIWTXVNPKYDL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium molybdate dihydrate (SMD) is a molybdic acid disodium salt. Its toxic effect on the avian species, northern bobwhite quail has been investigated.[1] Its ability to inhibit corrosion of 6082 wrought aluminum alloy has been studied in NaCl solution of chlorosulfonic acid.[2]

Application

Sodium molybdate dihydrate may be used to:
  • Prepare the modified Luria broth for growing Methylobacterium extorquens AM1 cells.[3]
  • Prepare shuttle-like barium molybdate (BaMoO4) microstructures under microwave conditions.[4]
  • Synthesize nickel-molybdenum-zinc (NiMoZn) electrode.[5]

Features and Benefits

Meets A.C.S. reagent specifications.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interconversion between formate and hydrogen carbonate by tungsten-containing formate dehydrogenase-catalyzed mediated bioelectrocatalysis.
Sakai K, et al.
Sensing and Bio-Sensing Research, 5, 90-96 (2015)
Corrosion and wear of 6082 aluminum alloy.
Panagopoulos CN, et al.
Tribology International, 42(6), 886-889 (2009)
Microwave-assisted synthesis, characterization and photoluminescence of shuttle-like BaMoO4 microstructure.
Phuruangrat A, et al.
Materials Science-Poland, 33(3), 537-540 (2015)
Dietary toxicity of soluble and insoluble molybdenum to northern bobwhite quail (Colinus virginianus).
Stafford JM, et al.
Ecotoxicology (London, England), 1-11 (2015)
Photoelectrochemical water splitting in an organic artificial leaf.
Esiner S, et al.
Journal of Material Chemistry A, 3(47), 23936-23945 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service