Skip to Content
Merck
All Photos(1)

Key Documents

32221

Sigma-Aldrich

Ethyl alcohol, pure

≥99.8% (GC), absolute, puriss. p.a., sales not in Germany

Synonym(s):

Ethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07
Beilstein:
1718733
EC Number:
MDL number:
UNSPSC Code:
15101802
PubChem Substance ID:

product name

Ethanol, puriss. p.a., absolute, ≥99.8% (GC)

vapor density

1.59 (vs air)

Quality Level

vapor pressure

44.6 mmHg ( 20 °C)

grade

absolute
puriss. p.a.

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

683 °F

quality

sales not in Germany1

expl. lim.

19 %, 60 °F

impurities

furfural, in accordance
≤0.0001% free alkali (as NH3)
≤0.0001% heavy metals (as Pb)
≤0.0003% KMnO4 red. matter (as O)
≤0.0005% free acid (as CH3COOH)
≤0.001% acetone (GC)
≤0.001% aldehydes (as CH3CHO)
≤0.001% non-volatile matter
≤0.003% 2-propanol
≤0.003% carbonyl compounds (as CO)
≤0.02% methanol (GC)
≤0.05% iso-amyl alcohol
≤0.2% water (Karl Fischer)

refractive index

n20/D 1.3600 (lit.)
n20/D 1.3612-1.3618

bp

78 °C (lit.)

mp

−114 °C (lit.)

density

0.789 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
B: ≤0.02 mg/kg
Ba: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sn: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

CCO

suitability

complies for UV absorption
complies for appearance
complies for identity A
complies for identity B
passes test for reaction against H2SO4

format

neat

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethanol was used as dispersion medium for already prepared silver nanoparticles.
It may be used in the following studies:
  • Hydrothermal synthesis of single crystal ZnO nanorod.
  • Synthesis of N-(N′dodecylcarboxamidomethyl)N,N,dimethyl ammonio propane sulfonates.
  • As solvent for cytofluorimetric analysis.
  • Preparation of TiO2 colloid (particle size = 20nm).

Footnote

1Distribution and use of ethyl alcohol in Switzerland require a license from the Swiss Federal Alcohol Administration

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qini Gan et al.
Journal of Alzheimer's disease : JAD, 70(4), 1025-1040 (2019-07-16)
Recent studies demonstrate that peripheral amylin treatment reduces pathology in mouse models of Alzheimer's disease (AD). However, soluble and aggregated amylin are distinct species; while amylin is a physiological neuropeptide, amylin aggregation is a pathological factor for diabetes. We thus
Francesca Filon Larese et al.
Toxicology, 255(1-2), 33-37 (2008-11-01)
There is a growing interest on nanoparticle safety for topical use. The benefits of nanoparticles have been shown in several scientific fields, but little is known about their potential to penetrate the skin. This study aims at evaluating in vitro
Synthesis of ZnO nanorod and the annealing effect on its photoluminescence property.
Wu L, et al.
Opt. Mater., 28(4), 418-422 (2006)
Christophe Tastet et al.
Proteomics, 3(2), 111-121 (2003-02-26)
Several zwitterionic detergents differing in their polar heads, linker parts and hydrophobia tail were synthesized and evaluated for their efficiency in protein solubilizers for two-dimensional electrophoresis. A model system consisting of human red blood cell ghosts was used for this
Fabrication of thin film dye sensitized solar cells with solar to electric power conversion efficiency over 10%.
Ito S, et al.
Thin Solid Films, 516(14), 4613-4619 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service