Skip to Content
Merck
All Photos(3)

Key Documents

206911

Sigma-Aldrich

Aluminum chloride

reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AlCl3
CAS Number:
Molecular Weight:
133.34
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38120507
PubChem Substance ID:
NACRES:
NA.21
Assay:
98%
grade:
reagent grade
form:
powder or chunks
Pricing and availability is not currently available.

grade

reagent grade

Quality Level

vapor pressure

1 mmHg ( 100 °C)

Assay

98%

form

powder or chunks

reaction suitability

reagent type: catalyst
core: aluminum

pH

2.4 (20 °C, 100 g/L)

mp

190 °C (lit.)

SMILES string

Cl[Al](Cl)Cl

InChI

1S/Al.3ClH/h;3*1H/q+3;;;/p-3

InChI key

VSCWAEJMTAWNJL-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Aluminum chloride participates in the amination of exo-2-chloronorbornane to afford 2-azabicyclo[3.2.1]octane, small amounts of the 3-isomer and exo-2-aminonorbornane.[1] It has been reported to promote the Friedel-Crafts acylation of 4-tert-butylbenzoyl chloride with mesitylene.[2]

Application

Aluminum chloride has been employed for the estimation of total contents of flavonoids in a freeze-dried extract isolated from Issopus officinalis.[3] It has been employed for the synthesis of the following compounds:[4]
  • 3′-methyl-5′,6′,7′,7a′-tetrahydro-4′H-spiro(cyclohexane-1,1′-isobenzofuran)
  • trans-methyl 2-(cyclohex-1-enyl)cyclohexyl ketone
  • 1,1,3-trimethyl-1,4,5,6,7,7a-hexahydroisobenzofuran
  • 1-((2R*,4aS*,8aR*)-2-methyldecahydronaphthalen-4ayl)ethanone
Aluminum chloride may be used as catalyst for the Friedel-Crafts polymerizations of 4-(trifluoromethoxy)benzyl bromide.[5] It may be employed as catalyst for Fries rearrangement of phenyl isobutyrate at 140°C to afford a mixture of 2-hydroxy- and 4-hydroxy-isobutyrophenone.[6]
It may be used to catalyze:
  • Degradation of polystyrene to benzene.[7]
  • Carbonization of polycyclic aromatic hydrocarbons.[8]
  • Colorimetric microassay of hydroperoxides.[9]

Features and Benefits

The vapor-phase co-reductions with other metal halides by hydrogen results in finely divided intermetallics with applications as structural materials or compounds with useful thermoelectric, magnetic, and oxidation-resistance properties. Aluminum nitride can be synthesized from the elemnts with AlCl3 added to help facilitate crystallization.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catalytic degradation of polystyrene in the presence of aluminum chloride catalyst.
Nanbu H, et al.
Polymer Degradation and Stability, 19(1), 61-76 (1987)
Aliphatic C-H activation with aluminium trichloride-acetyl chloride: expanding the scope of the Baddeley reaction for the functionalisation of saturated hydrocarbons.
Lyall CL, et al.
Organic & Biomolecular Chemistry, 11(9), 1468-1475 (2013)
The obtaining of an antioxidant product based on Issopus officinalis freeze: Dried extract.
Emanuel V, et al.
Journal of Medicinal Plants Research, 5(10), 2078-2085 (2011)
A colorimetric microdetermination of peroxide values utilizing aluminum chloride as the catalyst.
Asakawa T and Matsushita S.
Lipids, 15(11), 965-967 (1980)
Carbonization of pitches-IV: Carbonization of polycyclic aromatic hydrocarbons under the presence of aluminum chloride catalyst.
Mochida I, et al.
Carbon, 13(2), 135-139 (1975)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service