Y0001506
Lopinavir for peak identification
European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Lopinavir
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
lopinavir
manufacturer/tradename
EDQM
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
N4(CCCNC4=O)[C@@H](C(C)C)C(=O)N[C@H](C[C@H](O)[C@@H](NC(=O)COc3c(cccc3C)C)Cc2ccccc2)Cc1ccccc1
InChI
1S/C37H48N4O5/c1-25(2)34(41-20-12-19-38-37(41)45)36(44)39-30(21-28-15-7-5-8-16-28)23-32(42)31(22-29-17-9-6-10-18-29)40-33(43)24-46-35-26(3)13-11-14-27(35)4/h5-11,13-18,25,30-32,34,42H,12,19-24H2,1-4H3,(H,38,45)(H,39,44)(H,40,43)/t30-,31-,32-,34-/m0/s1
InChI key
KJHKTHWMRKYKJE-SUGCFTRWSA-N
Looking for similar products? Visit Product Comparison Guide
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Packaging
Other Notes
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
Lopinavir assay setup saves time and improves efficiency with shorter columns, meeting European pharmacopoeia guidelines.
Lopinavir assay setup saves time and improves efficiency with shorter columns, meeting European pharmacopoeia guidelines.
Lopinavir assay setup saves time and improves efficiency with shorter columns, meeting European pharmacopoeia guidelines.
Lopinavir assay setup saves time and improves efficiency with shorter columns, meeting European pharmacopoeia guidelines.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service