Skip to Content
Merck
All Photos(1)

Key Documents

Y0000416

Ranitidine impurity J

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1,1′-N-[Methylenebis(sulphanediylethylene)]bis(N′-methyl-2-nitroethene-1,1-diamine)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H22N6O4S2
CAS Number:
Molecular Weight:
366.46
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

ranitidine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

S(CSCCNC(=C[N+](=O)[O-])NC)CCNC(=C[N+](=O)[O-])NC

InChI

1S/C11H22N6O4S2/c1-12-10(7-16(18)19)14-3-5-22-9-23-6-4-15-11(13-2)8-17(20)21/h7-8,12-15H,3-6,9H2,1-2H3

InChI key

VWZXRGJMLLSQBK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ranitidine impurity J EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

  1. Y0000416 -RANTIDINE IMPURITY J Q1) QUANTITY PER PACK ? Q2) ARE THE EQUIVALENT WITH IPRS ?

    1 answer
    1. This is a product of the European Pharmacopoeia. There does not appear to be full harmonization of EP reference materials with the Indian Pharmacopoeia at this time. This item may not be an equivalent option recognized by the IPRS. Please see the link below to review a publication that may be helpful.
      https://www.edqm.eu/en/pharmacopoeial-harmonisation

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service