Skip to Content
Merck
All Photos(1)

Key Documents

Y0000173

4-Aminobenzoic acid

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

PABA, Vitamin Bx, Vitamin H1

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4CO2H
CAS Number:
Molecular Weight:
137.14
Beilstein:
471605
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

Agency

EP Reference Standard

API family

aminobenzoic acid

manufacturer/tradename

EDQM

mp

187-189 °C (lit.)

density

1.374 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Nc1ccc(cc1)C(O)=O

InChI

1S/C7H7NO2/c8-6-3-1-5(2-4-6)7(9)10/h1-4H,8H2,(H,9,10)

InChI key

ALYNCZNDIQEVRV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

4-Aminobenzoic acid EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

339.8 °F - closed cup

Flash Point(C)

171 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daisuke Koma et al.
Bioscience, biotechnology, and biochemistry, 78(2), 350-357 (2014-07-19)
The production of chemical compounds from renewable resources is an important issue in building a sustainable society. In this study, Escherichia coli was metabolically engineered by introducing T7lac promoter-controlled aroF(fbr), pabA, pabB, and pabC genes into the chromosome to overproduce
Heidi C Schilter et al.
Respiratory research, 16, 42-42 (2015-04-19)
The persistent influx of neutrophils into the lung and subsequent tissue damage are characteristics of COPD, cystic fibrosis and acute lung inflammation. VAP-1/SSAO is an endothelial bound adhesion molecule with amine oxidase activity that is reported to be involved in
Gerald H Jordan et al.
BJU international, 114(1), 16-24 (2014-01-23)
Peyronie's disease (PD) is often physically and psychologically devastating for patients, and the goal of treatment is to improve symptoms and sexual function without adding treatment-related morbidity. The potential for treatment-related morbidity after more invasive interventions, e.g. surgery, creates a
Kenshi Sankoda et al.
Archives of environmental contamination and toxicology, 68(2), 217-224 (2014-12-07)
This study aimed to investigate the behavior of organic ultraviolet (UV) filters released by recreational activities along the Japanese coastline. Seasonal variations of organic UV filters in seawater were investigated at four different recreational beaches (Mogushi, Wakamiya, Tsurugahama, and Otachimisaki
Xhorxhi Gjoka et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 972, 48-52 (2014-10-14)
Quantification of monoclonal antibody (mAb) monomer, mAb aggregates, and host cell proteins (HCPs) is critical for the optimization of the mAb production process. The present work describes a single high throughput analytical tool capable of tracking the concentration of mAb

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service