Skip to Content
Merck
All Photos(1)

Key Documents

Y0000146

Nadolol

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H27NO4
CAS Number:
Molecular Weight:
309.40
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

nadolol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(C)(C)NCC(O)COc1cccc2C[C@@H](O)[C@@H](O)Cc12

InChI

1S/C17H27NO4/c1-17(2,3)18-9-12(19)10-22-16-6-4-5-11-7-14(20)15(21)8-13(11)16/h4-6,12,14-15,18-21H,7-10H2,1-3H3/t12?,14-,15+/m1/s1

InChI key

VWPOSFSPZNDTMJ-UCWKZMIHSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Nadolol EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R C Heel et al.
Drugs, 20(1), 1-23 (1980-07-01)
Nadolol is a nonselective beta-adrenoceptor blocking drug without intrinsic sympathomimetic or membrane stabilising activity. Its inherently long duration of activity makes it suitable for once daily administration in both hypertension and angina pectoris, and in these conditions it has been
Nadolol: a new beta-adrenoceptor antagonist.
W H Frishman
The New England journal of medicine, 305(12), 678-682 (1981-09-17)
Zsuzsa Jenei-Lanzl et al.
Arthritis & rheumatology (Hoboken, N.J.), 66(9), 2472-2481 (2014-05-14)
Mesenchymal progenitor cell chondrogenesis is the biologic platform for the generation or regeneration of cartilage, but the external influence of the sympathetic nervous system on this process is not yet known. Sympathetic nerve fibers are present in articular tissue, and
S Misaka et al.
Clinical pharmacology and therapeutics, 95(4), 432-438 (2014-01-15)
This study aimed to evaluate the effects of green tea on the pharmacokinetics and pharmacodynamics of the β-blocker nadolol. Ten healthy volunteers received a single oral dose of 30 mg nadolol with green tea or water after repeated consumption of green
Rosa Adriana Jarillo-Luna et al.
Journal of neuroimmunology, 278, 223-231 (2014-12-04)
Stress stimuli affect the immune system of the mucosa, and in particular IgA secretion. It is well documented that intense psychological and physical stress can increase susceptibility to infection by diverse pathogens in the upper respiratory tract. Our workgroup reported

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service