Skip to Content
Merck
All Photos(1)

Key Documents

X102

Sigma-Aldrich

Triton X-102

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.21

description

non-ionic

Quality Level

mol wt

757 g/mol

CMC

0.33 mM

transition temp

cloud point 88 °C

HLB

14.4

SMILES string

O(CCOc1ccc(cc1)C(CC(C)(C)C)(C)C)CCO

InChI

1S/C18H30O3/c1-17(2,3)14-18(4,5)15-6-8-16(9-7-15)21-13-12-20-11-10-19/h6-9,19H,10-14H2,1-5H3

InChI key

LBCZOTMMGHGTPH-UHFFFAOYSA-N

Application


  • Effect of nonionic surfactants on the solubilization of alachlor.: This study explores the use of Triton X-102 in enhancing the solubility of alachlor, a widely used herbicide, demonstrating its potential to improve the environmental behavior and efficacy of agricultural chemicals (Xiarchos & Doulia, 2006).

  • Enantioselectivity of structurally modified poly(sodium undecenoyl-L-leucinate) by insertion of Triton X-102 surfactant molecules.: Investigates the modification of chiral separation media with Triton X-102, enhancing enantioselective capabilities in electrophoresis applications, critical for the pharmaceutical industry (Tarus et al., 2004).

  • Purification and characterization of human leukotriene C4 synthase.: Demonstrates the use of Triton X-102 in the purification processes of key enzymes involved in inflammatory responses, aiding in the development of anti-inflammatory drugs (Penrose et al., 1992).

  • Partial purification and characterization of erythropoietin receptors from erythroid progenitor cells.: Utilizes Triton X-102 to isolate and study erythropoietin receptors, contributing to better understanding and treatment of anemias (Im et al., 1990).

  • Isolation and characterization of insulin receptors from rat kidney glomeruli and tubules.: Explores the role of Triton X-102 in isolating specific receptors, enhancing knowledge on kidney function and potential targets for diabetes treatment (Im et al., 1988).


Legal Information

Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Colin E Moore et al.
ChemSusChem, 10(17), 3512-3519 (2017-07-01)
Bimetallic Sn-Pb catalysts with five different Sn/Pb atomic ratios were electrodeposited on Teflonated carbon paper and non-Teflonated carbon cloth using both fluoroborate- and oxide-containing deposition media to produce catalysts for the electrochemical reduction of CO
Haozhen Ren et al.
Liver international : official journal of the International Association for the Study of the Liver, 33(3), 448-458 (2013-01-11)
Hepatic tissue engineering is considered as a possible alternative to liver transplantation for end-stage liver disease. Several methods of decellularization of xenogeneic liver are available to produce three-dimensional organ scaffolds for engineering liver tissues. However, rare studies have examined and
Pil Seok Chae et al.
Molecular bioSystems, 9(4), 626-629 (2013-02-05)
Membrane protein manipulation is a challenging task owing to limited tertiary and quaternary structural stability once the protein has been removed from a lipid bilayer. Such instability can be overcome by embedding membrane proteins in detergent micelles formed from amphiphiles
Hirokazu Yokoyama et al.
Langmuir : the ACS journal of surfaces and colloids, 29(3), 857-860 (2012-12-29)
In this report, we present a novel approach for the elucidation of the physicochemical properties of lipid membranes by isothermal titration calorimetry (ITC) to quantify the heat absorbed during the solubilization of vesicles into TritonX-100 micelles. By using large and
Yonghui Wang et al.
Journal of the science of food and agriculture, 93(13), 3339-3344 (2013-04-19)
The production of pigments by Monascus spp. has attracted increasing attention. Modification of the cell membrane structure by addition of surfactants has proved to be effective for the secretion of intracellular metabolites. Hence in this study the effects and underlying

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service