Skip to Content
Merck
All Photos(1)

Key Documents

SMB00982

Sigma-Aldrich

L-Leucyl-L-phenylalanine

Synonym(s):

N-L-Leucyl-L-phenylalanine, N-Leucyl-3-phenylalanine, N-Leucylphenylalanine, L-Leu-L-Phe, Leu-phe, Leucine phenylalanine dipeptide, Leucyl-phenylalanine, Leucylphenylalanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22N2O3
CAS Number:
Molecular Weight:
278.35
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.26

Assay

≥95% (HPLC)

Quality Level

form

solid

mp

218—220 °C

storage temp.

2-8°C

SMILES string

[N+H3]C(CC(C)C)C(=O)NC(Cc1ccccc1)C(=O)[O-]

InChI

1S/C15H22N2O3/c1-10(2)8-12(16)14(18)17-13(15(19)20)9-11-6-4-3-5-7-11/h3-7,10,12-13H,8-9,16H2,1-2H3,(H,17,18)(H,19,20)

InChI key

KFKWRHQBZQICHA-UHFFFAOYSA-N

General description

L-Leucyl-L-phenylalanine, a dipeptide, is an incomplete breakdown product of protein digestion or protein catabolism.

Application

ʟ-Leucyl-ʟ-phenylalanine can be used in biochemical and metabolomics research applications

Features and Benefits

  • Suitable for Metabolomics and Biochemical research
  • High-quality compound suitable for multiple research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dustin G Brown et al.
Cancer & metabolism, 4, 11-11 (2016-06-09)
Colorectal cancers (CRC) are associated with perturbations in cellular amino acids, nucleotides, pentose-phosphate pathway carbohydrates, and glycolytic, gluconeogenic, and tricarboxylic acid intermediates. A non-targeted global metabolome approach was utilized for exploring human CRC, adjacent mucosa, and stool. In this pilot

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service