Skip to Content
Merck
All Photos(1)

Key Documents

S2050000

Sulfamethizole

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4-Amino-N-(5-methyl-1,3,4-thiadiazol-2-yl)benzenesulfonamide, N1-(5-Methyl-1,3,4-thiadiazol-2-yl)sulfanilamide, Sulfamethylthiadiazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10N4O2S2
CAS Number:
Molecular Weight:
270.33
Beilstein:
255002
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

sulfamethizole

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cc1nnc(NS(=O)(=O)c2ccc(N)cc2)s1

InChI

1S/C9H10N4O2S2/c1-6-11-12-9(16-6)13-17(14,15)8-4-2-7(10)3-5-8/h2-5H,10H2,1H3,(H,12,13)

InChI key

VACCAVUAMIDAGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sulfamethizole is a sulfonamide derivative used as an antimicrobial drug for the prevention and cure of bacterial infections in both humans and animals.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

This European Pharmacopoeia reference standard is intended for use only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solubility of sulfamethizole in some propylene glycol+ water mixtures at several temperatures.
Delgado D R, et al.
Fluid Phase Equilibria, 322, 113-119 (2012)
Susceptibility of Danish Escherichia coli strains isolated from urinary tract infections and bacteraemia, and distribution of sul genes conferring sulphonamide resistance.
Kerrn M B, et al.
The Journal of Antimicrobial Chemotherapy, 50(4), 513-516 (2002)
Adrian I Campos et al.
Molecular cell, 74(6), 1291-1303 (2019-05-03)
Alternative to the conventional search for single-target, single-compound treatments, combination therapies can open entirely new opportunities to fight antibiotic resistance. However, combinatorial complexity prohibits experimental testing of drug combinations on a large scale, and methods to rationally design combination therapies
Images in clinical medicine. Actinomycetoma.
Manuelle Viguier et al.
The New England journal of medicine, 372(3), 264-264 (2015-01-15)
Alexandre Grand-Guillaume Perrenoud et al.
Journal of chromatography. A, 1360, 275-287 (2014-08-19)
Superficially porous particles (SPP), or core shell particles, which consist of a non-porous silica core surrounded by a thin shell of porous silica, have gained popularity as a solid support for chromatography over the last decade. In the present study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service