PHR2103
Chlorphenamine Impurity D
Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
α-(4-Chlorophenyl)-α-[2-(dimethylamino)ethyl]-2-pyridineacetonitrile hydrochloride, (2RS)-2-(4-Chlorophenyl)-4-dimethylamino-2-(2-pyridyl)butyronitril hydrochloride, Chlorpheniramine nitrile hydrochloride
About This Item
Recommended Products
grade
certified reference material
pharmaceutical secondary standard
Quality Level
API family
chlorpheniramine
CofA
current certificate can be downloaded
packaging
pkg of 100 mg
application(s)
pharmaceutical small molecule
format
neat
storage temp.
2-8°C
SMILES string
ClC(C=C1)=CC=C1C(CCN(C)C)(C#N)C2=NC=CC=C2.Cl
InChI
1S/C17H18ClN3.ClH/c1-21(2)12-10-17(13-19,16-5-3-4-11-20-16)14-6-8-15(18)9-7-14;/h3-9,11H,10,12H2,1-2H3;1H
InChI key
PMJQCXXIWRUILE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Analysis Note
Other Notes
Footnote
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service