Skip to Content
Merck
All Photos(1)

Key Documents

PHR1939

Supelco

Hydroxyproline

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

trans-4-Hydroxy-L-proline, (2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid, Hyp

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO3
CAS Number:
Molecular Weight:
131.13
Beilstein:
81441
EC Number:
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

API family

proline

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

mp

273 °C (dec.) (lit.)

application(s)

pharmaceutical

format

neat

storage temp.

2-30°C

SMILES string

O[C@H]1CN[C@@H](C1)C(O)=O

InChI

1S/C5H9NO3/c7-3-1-4(5(8)9)6-2-3/h3-4,6-7H,1-2H2,(H,8,9)/t3-,4+/m1/s1

InChI key

PMMYEEVYMWASQN-DMTCNVIQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Hydroxyproline, found exclusively in collagen, is formed from the hydroxylation of proline. It is used as a biomarker to measure the collagen content of various biological samples.

Application

This pharmaceutical secondary standard can also be used as follows:

  • Multi-analysis of acetylcarnitine, proline, hydroxyproline, citrulline, and arginine as possible biomarkers by liquid chromatography-tandem mass spectrometry (LC-MS/MS) in human plasma samples
  • Simultaneous determination of proline and hydroxyproline in plasma samples by (LC-MS/MS using a new fluorescent reagent for pre-column derivatization
  • Development of a micellar electrokinetic chromatography (MEKC) method to measure hydroxyproline in various dairy products upon in-capillary derivatization
  • Quantitative analysis of free prolyl dipeptides, proline, and hydroxyproline by flow-gated capillary electrophoresis combined with laser-induced fluorescence detector from unhydrolyzed urine samples[1]

Other Notes

Natural constituent of animal structural proteins such as collagen and elastin.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC2898 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ning Zhang et al.
Analytical and bioanalytical chemistry, 409(30), 7077-7085 (2017-10-04)
Unhydrolyzed prolyl hydroxyproline (Pro-Hyp) and total 4-hydroxyproline (Hyp) in urine have been suggested as disease biomarkers for bone turnover and osteoporosis. Here, a rapid method was developed to accurately and selectively determine free prolyl compounds in unhydrolyzed urine samples. Urine

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. Does the data indicate that refrigeration temperatures (2-8°C) or freezing would help maintain the stability of the amino acid standard? Additionally, is it recommended to store the powder in a desiccator to preserve its stability over time?

    1 answer
    1. There have been no indications of instability in this material in either short-term or long-term studies. Storing the material at cooler temperatures would help minimize the risk of degradation. However, it's important to note that cannot guarantee the ongoing suitability of a standard over multiple uses.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service