Skip to Content
Merck
All Photos(1)

Key Documents

PHR1891

Supelco

Tolterodine Tartrate

Pharmaceutical Secondary Standard: Certified Reference Material

Synonym(s):

(R)-Tolterodine L-tartrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H31NO · C4H6O6
CAS Number:
Molecular Weight:
475.57
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0001696
traceable to USP 1671480

API family

tolterodine

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

application(s)

pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

N(C(C)C)(C(C)C)CC[C@H](c2c(ccc(c2)C)O)c1ccccc1.O[C@H]([C@@H](O)C(=O)O)C(=O)O

InChI

1S/C22H31NO.C4H6O6/c1-16(2)23(17(3)4)14-13-20(19-9-7-6-8-10-19)21-15-18(5)11-12-22(21)24;5-1(3(7)8)2(6)4(9)10/h6-12,15-17,20,24H,13-14H2,1-5H3;1-2,5-6H,(H,7,8)(H,9,10)/t20-;1-,2-/m01/s1

InChI key

TWHNMSJGYKMTRB-CDHDNNKHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tolterodine Tartrate is a tertiary muscarinic antagonist, basically used to manage urinary frequency, urgency, and incontinence in detrusor instability.[1]

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.
Tolterodine Tartrate may be used as a pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations using spectrophotometric technique.[1]

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB0501 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of tolterodine tartrate in pharmaceutical preparations using eosin, application to stability study
Walash IM, et al.
International Journal of Pharmaceutical Sciences and Research, 2(11), 2849-2849 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service