PHR1824
Pramipexole Related Compound D
Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Dexpramipexole dihydrochloride, (R)-2-Amino-4,5,6,7-tetrahydro-6-(propylamino)benzothiazole dihydrochloride, (R)-2-Amino-4,5,6,7-tetrahydro-6-(propylamino)benzothiazole dihydrochloride // (R)-Pramipexole dihydrochloride, (R)-4,5,6,7-Tetrahydro-6-(propylamino)-benzothiazole-2-amine dihydrochloride, (R)-Pramipexole dihydrochloride, KNS 760704, R-(+)-Pramipexole dihydrochloride, RPPX, SND 919CL2X
About This Item
Recommended Products
grade
certified reference material
pharmaceutical secondary standard
Quality Level
Agency
traceable to Ph. Eur. Y0001267
traceable to USP 1553815
description
Pharmaceutical Secondary Standard; Certified Reference Material
API family
pramipexole
CofA
current certificate can be downloaded
packaging
pkg of 50 mg
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-30°C
SMILES string
Cl.Cl.CCCN[C@@H]1CCc2nc(N)sc2C1
InChI
1S/C10H17N3S.2ClH/c1-2-5-12-7-3-4-8-9(6-7)14-10(11)13-8;;/h7,12H,2-6H2,1H3,(H2,11,13);2*1H/t7-;;/m1../s1
InChI key
QMNWXHSYPXQFSK-XCUBXKJBSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Biochem/physiol Actions
Analysis Note
Other Notes
Footnote
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service