Skip to Content
Merck
All Photos(1)

Key Documents

PHR1716

Supelco

Capreomycin Sulfate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Capreomycin sulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 59
traceable to USP 1091006

API family

capreomycin

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(O)O.N1C(CCNC1=N)C2NC(=O)\C(=C\NC(=O)N)\NC(=O)C(NC(=O)C(NC(=O)C(CNC2=O)N)CO)CNC(=O)CC(N)CCCN

InChI

1S/C25H44N14O8.H2O4S/c26-4-1-2-11(27)6-17(41)32-8-14-20(43)35-15(9-34-25(30)47)21(44)39-18(13-3-5-31-24(29)38-13)23(46)33-7-12(28)19(42)37-16(10-40)22(45)36-14;1-5(2,3)4/h9,11-14,16,18,40H,1-8,10,26-28H2,(H,32,41)(H,33,46)(H,35,43)(H,36,45)(H,37,42)(H,39,44)(H3,29,31,38)(H3,30,34,47);(H2,1,2,3,4)/b15-9-;

InChI key

AJQVUIHGEOLMDY-SOCRLDLMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Capreomycin sulfate is a disulfate salt of the polypeptide antibiotic drug, capreomycin, that is administered either parenterally or in the form of aerosols. It is generally used against multidrug-resistant tuberculosis (MDR-TB).
Pharmaceutical secondary standard for application in quality control. Provides pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Capreomycin sulfate may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3042 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhaled large porous particles of capreomycin for treatment of tuberculosis in a guinea pig model
Garcia-Contreras L, et al.
Antimicrobial Agents and Chemotherapy, 51(8), 2830-2836 (2007)
Development and validation of a liquid chromatographic method for the analysis of capreomycin sulfate and its related substances
Mallampati S, et al.
Journal of Chromatography A, 1216(12), 2449-2455 (2009)
Isolation and identification of four major impurities in capreomycin sulfate
Liu G, et al.
Journal of Chromatography A, 1571(12), 155-164 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service