Skip to Content
Merck
All Photos(1)

Key Documents

PHR1265

Supelco

Carvedilol

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Carvedilol, 1-(9H-Carbazol-4-yloxy)-3-[[2-(2-methoxyphenoxy)ethyl]amino]-2-propanol, BM-14190

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H26N2O4
CAS Number:
Molecular Weight:
406.47
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0001427
traceable to USP 1096622

API family

carvedilol

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

OC(COC1=CC=CC2=C1C3=C(C=CC=C3)N2)CNCCOC4=CC=CC=C4OC

InChI

1S/C24H26N2O4/c1-28-21-10-4-5-11-22(21)29-14-13-25-15-17(27)16-30-23-12-6-9-20-24(23)18-7-2-3-8-19(18)26-20/h2-12,17,25-27H,13-16H2,1H3

InChI key

OGHNVEJMJSYVRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Carvedilol is a lipophilic β1- and β2-adrenoreceptor antagonist that exhibits antioxidant and antiproliferative properties. It is prescribed as a medicine for the treatment of hypertension, stable angina pectoris, post myocardial infarction with left ventricular dysfunction and all degrees of symptomatic chronic heart failure.

Application

Carvedilol may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations and serum samples by various chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Cavedilol is a non-selective β-adrenergic blocker with α1 blocking activity. Carvedilol is used specifically for the treatment of heart failure and high blood pressure. It has been shown to improve left ventricular ejection fraction and may reduce mortality.

Caution

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA9063 in the slot below. This is an example certificate only and may not be the lot that you receive.

related product

Product No.
Description
Pricing

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2 - STOT RE 2

Target Organs

Liver,spleen,Uterus (including cervix),Adrenal gland

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Achiral-chiral LC/LC-FLD coupling for determination of carvedilol in plasma samples for bioequivalence purposes
Medvedovici A, et al.
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 850(1-2), 327-335 (2007)
HPLC Method Development and Validation of S (-)-Carvedilol from API and Formulations
Swetha E, et al.
American Journal of Analytical Chemistry, 6(05), 437-437 (2015)
Simple and accurate RP-HPLC and TLC-Densitometric methods for determination of carvedilol in pharmaceutical formulations
Abdel-Gawad FM, et al.
International Journal of Research in Pharmacy and Chemistry, 2(3), 2231-2781 (2012)
Quantification of carvedilol in human plasma by high-performance liquid chromatography coupled to electrospray tandem mass spectrometry: application to bioequivalence study
do Carmo Borges NC, et al.
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 822(1-2), 253-262 (2005)
Yanzhuo Zhang et al.
International journal of pharmaceutics, 454(1), 403-411 (2013-07-16)
The main objective of this study was to develop carboxylated ordered mesoporous carbon microparticles (c-MCMs) loaded with a poorly water-soluble drug, intended to be orally administered, able to enhance the drug loading capacity and improve the oral bioavailability. A model

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service